missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.01N Volumetric Solution, BAKER ANALYZED™ Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

Manufacturer:  Avantor Performance 566303

 View more versions of this product

Catalog No. 02004101


 

Specifications

Specifications

Sodium Hydroxide, 0.01N Volumetric Solution
1310-73-2
HNaO
[OH-].[Na+]
39.997
CHEBI:32145
Cubetainer
4L
BAKER ANALYZED™ Reagent
HEMHJVSKTPXQMS-UHFFFAOYSA-M
sodium;hydroxide
14798
40
1L = 1.00kg

Chemical Identifiers

1310-73-2
39.997
14798
HNaO
HEMHJVSKTPXQMS-UHFFFAOYSA-M
CHEBI:32145
Documents