Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hydroxide, 0.01N Standardized Solution

Chemical Identifiers

Chemical Name or Material Sodium hydroxide
CAS 1310-73-2
Name Note Standardized Solution
Formula Weight 40
MDL Number MFCD00003548
Synonym Caustic soda; Sodium hydrate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA35620K2

 
alfa aesar™
35620K2
1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35620K7

 
alfa aesar™
35620K7
4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA35620M4

 
alfa aesar™
35620M4
20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 40
Chemical Name or Material Sodium hydroxide
TSCA TSCA
Quantity 1L
Name Note Standardized Solution
Physical Form Liquid
Concentration 0.01N
MDL Number MFCD00003548
CAS 1310-73-2
Synonym Caustic soda; Sodium hydrate
Odor Odorless