Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.0100N, APHA for Sulfide, Ricca Chemicals

$47.76 - $108.82

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2 (Sodium Hydroxide)
CAS 7732-18-5 (Water, Deionized)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7295-1

 
ricca chemical company
RI72951
Plastic 4L Each for $108.82

7295-32

 
ricca chemical company
RI729532
Plastic 1L Each for $47.76

7295-25

 
ricca chemical company
RI72952.5
Cubitainer 10L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 4L
Methods APHA for Sulfide
Chemical Name or Material Sodium Hydroxide
CAS 1310-73-2 (Sodium Hydroxide)
Concentration or Composition (by Analyte or Components) 0.0100N
Packaging Plastic
CAS 7732-18-5 (Water, Deionized)