Please login to your online account to display your discounted pricing

Sodium Hydroxide, 0.0100 Normal, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 1310-73-2
CAS Min % 0.04
CAS Max % 0.04
Physical Form Liquid
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

729516

 
ricca chemical company
RI729516
Poly natural 500mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

72955

 
ricca chemical company
RI72955
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R72950004F

 
ricca chemical company
RIR72950004F
Cubitainer 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Quantity 500mL
Chemical Name or Material Sodium Hydroxide
CAS 7732-18-5
CAS 1310-73-2
Concentration or Composition Notes 0.0100Normal
CAS Max % 0.04
CAS Min % 0.04
CAS Max % 100.0
Packaging Poly natural
CAS Min % 98.0