Learn More
Sodium hydrosulfite, ca. 85%, Tech., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 169590250
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Reduction reagentSpecifications
Sodium hydrosulfite | |
tech., ca. 85% | |
White | |
25 g | |
Na2O4S2 | |
MFCD00011640 | |
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros | |
JVBXVOWTABLYPX-UHFFFAOYSA-L | |
174.1 | |
CHEBI:66870 | |
∼85% | |
Glass bottle |
>300.0°C | |
7775-14-6 | |
Crystalline Powder | |
ca. 85% | |
Na2S2O4 | |
15, 8747 | |
Solubility in water: 250g/L (20°C). Other solubilities: slightly soluble in alcohol | |
[O-]S(=O)S(=O)[O-].[Na+].[Na+] | |
24489 | |
174.1 | |
Technical |
Chemical Identifiers
7775-14-6 | |
JVBXVOWTABLYPX-UHFFFAOYSA-L | |
24489 | |
[O-]S(=O)S(=O)[O-].[Na+].[Na+] |
MFCD00011640 | |
sodium dithionite, sodium hydrosulfite, disodium dithionite, vatrolite, sodium sulfoxylate, sodium hydrosulphite, dithionous acid, disodium salt, blankit, burmol, hydros | |
CHEBI:66870 |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Self-heating; may catch fire.
Contact with acids liberates toxic gas.
GHS P Statement
Keep cool.
Protect from sunlight.
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
GHS Signal Word: Danger
EINECSNumber : 231-890-0
TSCA : TSCA
RUO – Research Use Only