missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydrogencyanamide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438050250

 View more versions of this product

Catalog No. AC438050250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Sodium hydrogencyanamide
98%
cyanamide monosodium salt, cyanamide, monosodium salt, cyanamide, sodium salt 1:1, monosodium cyanamide, sodium acid cyanamide, sodium cyanamidate, sodium cyanamide, sodium cyanoazanide, sodium hydrogen cyanamide, sodium hydrogencyanamide
MBEGFNBBAVRKLK-UHFFFAOYSA-N
sodium;cyanoazanide
4293993
Crystalline Powder
Authentic
Solubility in water: 800g/L (20°C).
>360.0°C
17292-62-5
CHN2Na
Solubility in water: 800g/L (20°C)
C(#N)[NH-].[Na+]
64.02
64.02
98%
Glass bottle
Tan to White
25g

Chemical Identifiers

17292-62-5
64.02
cyanamide monosodium salt, cyanamide, monosodium salt, cyanamide, sodium salt 1:1, monosodium cyanamide, sodium acid cyanamide, sodium cyanamidate, sodium cyanamide, sodium cyanoazanide, sodium hydrogen cyanamide, sodium hydrogencyanamide
sodium;cyanoazanide
CHN2Na
MBEGFNBBAVRKLK-UHFFFAOYSA-N
4293993
C(#N)[NH-].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath

GHS Signal Word: Danger

EINECSNumber : 241-322-3

SDS Documents