missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hydrogencyanamide, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC438050050
Specifications
Sodium hydrogencyanamide | |
98% | |
cyanamide monosodium salt, cyanamide, monosodium salt, cyanamide, sodium salt 1:1, monosodium cyanamide, sodium acid cyanamide, sodium cyanamidate, sodium cyanamide, sodium cyanoazanide, sodium hydrogen cyanamide, sodium hydrogencyanamide | |
MBEGFNBBAVRKLK-UHFFFAOYSA-N | |
sodium;cyanoazanide | |
4293993 | |
Crystalline Powder | |
Authentic | |
Solubility in water: 800g/L (20°C). | |
>360.0°C |
17292-62-5 | |
CHN2Na | |
Solubility in water: 800g/L (20°C) | |
C(#N)[NH-].[Na+] | |
64.02 | |
64.02 | |
98% | |
Glass bottle | |
Tan to White | |
5g |
Chemical Identifiers
17292-62-5 | |
64.02 | |
cyanamide monosodium salt, cyanamide, monosodium salt, cyanamide, sodium salt 1:1, monosodium cyanamide, sodium acid cyanamide, sodium cyanamidate, sodium cyanamide, sodium cyanoazanide, sodium hydrogen cyanamide, sodium hydrogencyanamide | |
sodium;cyanoazanide |
CHN2Na | |
MBEGFNBBAVRKLK-UHFFFAOYSA-N | |
4293993 | |
C(#N)[NH-].[Na+] |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breath
GHS Signal Word: Danger
EINECSNumber : 241-322-3