Please login to your online account to display your discounted pricing

Sodium hydrogencyanamide, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Sodium hydrogencyanamide
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula CHN2Na
Formula Weight 64.02
Synonym Cyanamide monosodium salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC438050050

  SDS
thermo fisher scientific
AC438050050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC438050250

  SDS
thermo fisher scientific
AC438050250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 64.02
Quantity 5g
Color White to tan
Chemical Name or Material Sodium hydrogencyanamide
Infrared Spectrum Authentic
Synonym Cyanamide monosodium salt
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula CHN2Na
CAS 17292-62-5
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.