missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydrogen sulfide, Anhydrous, Alfa Aesar™

For Use in Paper Pulping, in Dyestuffs Processing, in Rayon and Cellophane Desulfurizing

Manufacturer:  Alfa Aesar 8738904

 View more versions of this product

Catalog No. AA8738904



Description

Description

Moisture Sensitive and Hygroscopic
Specifications

Specifications

Sodium hydrogen sulfide
HNaS
sodium hydrosulfide, sodium bisulfide, sodium sulfhydrate, sodium hydrogen sulfide, sodium sulfide na sh, sodium sulfanide, sodium mercaptan, sodium mercaptide, sodium hydrosulphide, nahs
[Na+].[SH-]
56.058
56.06
1.79g/cm3 at 20°C
Moisture sensitive
White
16721-80-5
MFCD00011124
HYHCSLBZRBJJCH-UHFFFAOYSA-M
sodium;sulfanide
28015
Crystalline
Like rotten eggs
Anhydrous
2g

Chemical Identifiers

16721-80-5
56.058
HYHCSLBZRBJJCH-UHFFFAOYSA-M
28015
[Na+].[SH-]
HNaS
MFCD00011124
sodium hydrosulfide, sodium bisulfide, sodium sulfhydrate, sodium hydrogen sulfide, sodium sulfide na sh, sodium sulfanide, sodium mercaptan, sodium mercaptide, sodium hydrosulphide, nahs
sodium;sulfanide
Safety and Handling

Safety and Handling

Keep container in a well ventilated place. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.

H228-H251-H301-H311-H332-H314-H318: Flammable solid. Self-heating: may catch fire. Toxic if swallowed. Toxic in contact with skin. Harmful if inhaled. Causes severe skin burns and eye damage. Causes serious eye damage.

DOTInformation : DOT Class: 4.2, Packaging Group: II

EINECSNumber : 240-778-0

SDS Documents