missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hydrogen sulfate, anhydrous, 90+%, remainder mainly sodium sulfate, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B255870B
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hydrogen sulfate | |
anhydrous | |
1.0 | |
remainder mainly sodium sulfate | |
MFCD00003531 | |
14,8586 | |
WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
sodium;hydrogen sulfate | |
516919 | |
≥90% | |
Hygroscopic |
177°C to 180°C | |
7681-38-1 | |
1000 g | |
HNaO4S | |
UN3260 | |
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
[Na+].OS([O-])(=O)=O | |
120.05 | |
120.06 | |
Odorless | |
2.435 |
Chemical Identifiers
7681-38-1 | |
120.05 | |
WBHQBSYUUJJSRZ-UHFFFAOYSA-M | |
516919 |
HNaO4S | |
MFCD00003531 | |
sodium hydrogen sulfate, sodium bisulfate, sodium acid sulfate, bisulfate of soda, sodium hydrosulfate, fanal, sodium hydrogensulphate, niter cake, nitre cake, wc-perfect | |
[Na+].OS([O-])(=O)=O |
Safety and Handling
GHS H Statement
H318
Causes serious eye damage.
P280-P305+P351+P338-P310
H318
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber : 231-665-7
RTECSNumber : VZ1860000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only