missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydrogen phosphate heptahydrate, ACS, 98.0-102.0%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1159236

 View more versions of this product

Catalog No. AA1159236



Specifications

Specifications

Sodium hydrogen phosphate heptahydrate
98.0-102.0%
MFCD00149180
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
268.062
268.07; 141.98 (anhydrous)
ACS
Odorless
500g
7782-85-6
H15Na2O11P
White crystals
PYLIXCKOHOHGKQ-UHFFFAOYSA-L
disodium;hydrogen phosphate;heptahydrate
6096963
Crystalline
1.7g/cm3 at 20°C
White

Chemical Identifiers

7782-85-6
268.062
PYLIXCKOHOHGKQ-UHFFFAOYSA-L
6096963
O.O.O.O.O.O.O.OP(=O)([O-])[O-].[Na+].[Na+]
H15Na2O11P
MFCD00149180
sodium phosphate dibasic heptahydrate, unii-70wt22sf4b, sodium phosphate, dibasic, heptahydrate, sodium hydrogen phosphate heptahydrate, disodium hydrogen phosphate heptahydrate, axea paragraph signthornaae, 2na.hpo4.7h2o, sodium hydrogenphosphate heptahydrate, sodium phosphate dibasic, acs, disodium hydrogenphosphate heptahydrate
disodium;hydrogen phosphate;heptahydrate
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 231-448-7

Documents