missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydrogen phosphate, anhydrous, ACS, 99.0% min., Alfa Aesar™

Manufacturer:  Alfa Aesar 13437A1

 View more versions of this product

Catalog No. AA13437A1



Specifications

Specifications

Sodium hydrogen phosphate, anhydrous
99.0% min.
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
OP(=O)([O-])[O-].[Na+].[Na+]
141.957
CHEBI:34683
Granules
Maximum level of impurities: Insoluble matter 0.01%, Loss on drying at 105° 0.2%, pH of a 5% solution 8.7 to 9.3 at 25°, Cl 0.002%, SO4 0.005%, Heavy Metals (as Pb) 0.001%, Fe 0.002%
Anhydrous
1kg
7558-79-4
HNa2O4P
White granular
BNIILDVGGAEEIG-UHFFFAOYSA-L
disodium;hydrogen phosphate
24203
141.98
ACS
Odorless
White

Chemical Identifiers

7558-79-4
141.957
BNIILDVGGAEEIG-UHFFFAOYSA-L
24203
disodium;hydrogen phosphate
HNa2O4P
MFCD00003496
disodium hydrogen phosphate, disodium phosphate, disodium hydrogenorthophosphate, sodium phosphate dibasic, dibasic sodium phosphate, acetest, soda phosphate, disodium orthophosphate, sodium hydrogen phosphate, phosphoric acid, disodium salt
CHEBI:34683
OP(=O)([O-])[O-].[Na+].[Na+]
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable gloves. This material and/or its container must be disposed of as hazardous waste.

H315-H319-H335: Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.

EINECSNumber : 231-448-7

Documents