missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hydrogen L-tartrate, anhydrous, 98%

Manufacturer:  Alfa Aesar™ L1246914

 View more versions of this product

Catalog No. AAL1246914



Description

Description

Applications
Sodium hydrogen L-tartrate is used for detecting potassium and in nutrient media. It also used as a food additive and an acidity regulator.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

Sodium hydrogen L-tartrate
~243°C
anhydrous
C4H5NaO6
6099125
sodium bitartrate, monosodium tartrate, sodium hydrogen tartrate, natriumtartrat german, sodium 3-carboxy-2,3-dihydroxypropanoate, monobasic sodium tartrate, monosodium l-+-tartrate, tartaric acid, monosodium salt, natrium hydrogen-2r,3r-tartrat, weinstein
C(C(C(=O)[O-])O)(C(=O)O)O.[Na+]
172.068
172.07
+24.5° (c=3 in Water)
25g
526-94-3
MFCD00065393
14,8589
NKAAEMMYHLFEFN-UHFFFAOYSA-M
sodium;2,3,4-trihydroxy-4-oxobutanoate
23690454
98%

Chemical Identifiers

526-94-3
172.068
NKAAEMMYHLFEFN-UHFFFAOYSA-M
23690454
C(C(C(=O)[O-])O)(C(=O)O)O.[Na+]
C4H5NaO6
MFCD00065393
sodium bitartrate, monosodium tartrate, sodium hydrogen tartrate, natriumtartrat german, sodium 3-carboxy-2,3-dihydroxypropanoate, monobasic sodium tartrate, monosodium l-+-tartrate, tartaric acid, monosodium salt, natrium hydrogen-2r,3r-tartrat, weinstein
sodium;2,3,4-trihydroxy-4-oxobutanoate
Safety and Handling

Safety and Handling

RTECSNumber : WW8225000

TSCA : TSCA

Documents