Learn More
Sodium hydrogen carbonate, Puratronic™, 99.998% (metals basis), Thermo Scientific Chemicals
m.p. approx. 50°C, begins to lose CO2 approx.100°C, converts to Na2CO3
Supplier: Thermo Scientific Chemicals 01086309
Description
Sodium hydrogen carbonate is used in a process for removing paint and corrosion called sodablasting; the process is particularly suitable for cleaning aluminum panels which can be distorted by other types of abrasive. It can be administered to pools, spas, and garden ponds to raise pH levels. Also can be used as a fire extinguisher. Its primary use is in cooking (baking), as a leavening agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hydrogen carbonate | |
White | |
10 g | |
CHNaO3 | |
4153970 | |
Soluble in water. Insoluble in ethanol. | |
[Na+].OC([O-])=O | |
84.01 | |
CHEBI:32139 | |
Odorless | |
White powder |
144-55-8 | |
Powder | |
99.998% (metals basis) | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
sodium hydrogen carbonate | |
516892 | |
84.01 | |
1.5 | |
2.159 |
Chemical Identifiers
144-55-8 | |
84.01 | |
UIIMBOGNXHQVGW-UHFFFAOYSA-M | |
516892 | |
[Na+].OC([O-])=O |
CHNaO3 | |
MFCD00003528 | |
sodium bicarbonate, sodium hydrogen carbonate, baking soda, carbonic acid monosodium salt, sodium acid carbonate, bicarbonate of soda, sodium hydrogencarbonate, meylon, acidosan, neut | |
CHEBI:32139 |
Safety and Handling
H303: May be harmful if swallowed.
EINECSNumber : 205-633-8
RTECSNumber : VZ0950000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only