missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydrogen carbonate, 99.998% (metals basis), Alfa Aesar™, Puratronic™

m.p. approx. 50°C, begins to lose CO2 approx.100°C, converts to Na2CO3

Manufacturer:  Thermo Fisher Scientific 1086318

 View more versions of this product

Catalog No. AA1086318



Specifications

Specifications

Sodium hydrogen carbonate
99.998% (metals basis)
MFCD00003528
acidosan
C(=O)(O)[O-].[Na+]
84.006
CHEBI:32139
Powder
Odorless
50g
144-55-8
CHNaO3
White powder
UIIMBOGNXHQVGW-UHFFFAOYSA-M
sodium;hydrogen carbonate
516892
84.01
2.159g/cm3 at 20°C
White

Chemical Identifiers

144-55-8
84.006
UIIMBOGNXHQVGW-UHFFFAOYSA-M
516892
sodium;hydrogen carbonate
CHNaO3
MFCD00003528
acidosan
CHEBI:32139
C(=O)(O)[O-].[Na+]
Safety and Handling

Safety and Handling

H303: May be harmful if swallowed.

EINECSNumber : 205-633-8

Documents