Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hydride, ca 60% dispersion in oil, in solvent solublebags

Chemical Identifiers

Chemical Name or Material Sodium hydride
CAS 7646-69-7
Name Note ≈60% dispersion in oil, in solvent solublebags
Formula Weight 24
MDL Number MFCD00003471
UN Number UN1427
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3649018

 
alfa aesar™
H3649018
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3649022

 
alfa aesar™
H3649022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH3649036

 
alfa aesar™
H3649036
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 24
Chemical Name or Material Sodium hydride
TSCA TSCA
Odor Odorless
Name Note ≈60% dispersion in oil, in solvent solublebags
Physical Form Moist Powder
Sensitivity Air & Moisture Sensitive
MDL Number MFCD00003471
CAS 7646-69-7
UN Number UN1427
Color White to pale gray
Quantity 50g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H260
In contact with water releases flammable gases which may ignite spontaneously.
Health Hazard 3 GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: SODIUM HYDRIDE

Applications
Sodium hydride can be used for direct intercalation into C60, resulting in superconducting material (NaH)4C60. It is primarily used as a strong base in organic synthesis. It can be used to dry some organic solvents. It has been proposed for hydrogen storage for use in fuel cell vehicles.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, acid chlorides, acids, halogens and alcohols.

Safety and Handling

Danger

GHS P Statement
P231+P232-P280-P233-P370+P378c-P402+P404-P501a
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
Keep container tightly closed.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H260
In contact with water releases flammable gases which may ignite spontaneously.