missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hydride, 60% dispersion in mineral oil, in soluble bags, ACROS Organics™

Manufacturer:  Acros Organics 189861000

 View more versions of this product

Catalog No. AC189861000


 

Description

Description

Adsorbent, Reduction reagent
Specifications

Specifications

Sodium hydride, 60% dispersion in mineral oil
57 to 63% (NaH)
NaH
02,382; 01,1075; 04,452; 05,610; 06,541; 07,335; 08,458; 09,427; 11,486; 12,447; 14,288; 16,307
Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
[H-].[Na+]
23.998
24
1.2000g/mL
Gray to Yellow
100g
7646-69-7,8042-47-5
HNa
MFCD00003471
15, 8760
BZKBCQXYZZXSCO-UHFFFAOYSA-N
sodium;hydride
24758
Tacky Powder
Solubility in water: reacts. Other solubilities: reacts with alcohols, soluble in molten sodium hydroxide, insoluble in cs2, ccl4, liquid nh3, benzene
800.0°C
165°C

Chemical Identifiers

7646-69-7
23.998
BZKBCQXYZZXSCO-UHFFFAOYSA-N
HNa
MFCD00003471
24758
Safety and Handling

Safety and Handling

GHS H Statement:
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement:


WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

SDS Documents