missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium hydride, 57-63% oil dispersion, Alfa Aesar™
Manufacturer: Alfa Aesar 1343136
Specifications
Sodium hydride | |
HNa | |
Off-white to gray moist powder | |
BZKBCQXYZZXSCO-UHFFFAOYSA-N | |
sodium;hydride | |
24758 | |
Moist Powder | |
White to Gray |
7646-69-7 | |
MFCD00003471 | |
sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid | |
[H-].[Na+] | |
23.998 | |
24 | |
Odorless | |
500g |
Chemical Identifiers
7646-69-7 | |
23.998 | |
BZKBCQXYZZXSCO-UHFFFAOYSA-N | |
24758 | |
[H-].[Na+] |
HNa | |
MFCD00003471 | |
sodium hydride, sodiumhydride, sodium hydride nah, sodium hydride oil dispersion, sodium monohydride, sodium hydride, sodium hydride nad, sodiumhydrid, natrium hydride, sodium hydrid | |
sodium;hydride |
Safety and Handling
H260: In contact with water releases flammable gases which may ignite spontaneously.
DOTInformation : Hazard Class: 4.3, Packing Group: I
EINECSNumber : 231-587-3