missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hippurate, 96%

Manufacturer:  Alfa Aesar A1172318

 View more versions of this product

Catalog No. AAA1172318


 

Description

Description

Applications
It may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Sodium Hippurate
96%
C9H8NNaO3
ZBCAZEFVTIBZJS-UHFFFAOYSA-M
sodium;2-benzamidoacetate
516953
Lump/Powder
532-94-5
96%
MFCD00002693
C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+]
201.157
201.16
50g

Chemical Identifiers

532-94-5
201.157
ZBCAZEFVTIBZJS-UHFFFAOYSA-M
C9H8NNaO3
MFCD00002693
516953
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS missing translation for 'documents'