missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hippurate, 96%, Alfa Aesar™

Manufacturer:  Alfa Aesar A1172330

 View more versions of this product

Catalog No. AAA1172330



Description

Description

Applications
It may be a useful urinary biomarker for certain cancers such as gastric cancer and oxidative stress.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications

Specifications

Sodium hippurate
532-94-5
C9H8NNaO3
sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt
C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+]
201.157
201.16
96%
250g
96%
MFCD00002693
ZBCAZEFVTIBZJS-UHFFFAOYSA-M
sodium;2-benzamidoacetate
516953
Lump/Powder

Chemical Identifiers

532-94-5
201.157
ZBCAZEFVTIBZJS-UHFFFAOYSA-M
516953
C1=CC=C(C=C1)C(=O)NCC(=O)[O-].[Na+]
C9H8NNaO3
MFCD00002693
sodium hippurate, hippuric acid sodium salt, sodium 2-benzamidoacetate, n-benzoylglycine sodium salt, glycine, n-benzoyl-, monosodium salt, ccris 5812, hippuric acid, monosodium salt, glycine, n-benzoyl-, sodium salt 1:1, hippurate sodium, benzoylaminoacetic acid sodium salt
sodium;2-benzamidoacetate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents