Please login to your online account to display your discounted pricing

Sodium hexanitrocobaltate(III), ACROS Organics™


Quantity 500g

Manufacturer: thermo fisher scientific  AC214865000

 View more versions of this product

Catalog No. AC214865000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Powder
Formula Weight 403.94
Quantity 500g
Color Orange to orange-brown
Synonym Sodium cobaltinitrite
Linear Formula Na3Co(NO2)6
Molecular Formula CoN6Na3O12
Chemical Name or Material Sodium hexanitrocobaltate(III)
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Immerse in cool water/wrap in wet bandages. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Suitability (for determination of potassium) passes
Insoluble Matter 0.02% max. (in dil. CH3COOH)
CAS 13600-98-1
MDL Number MFCD00003512
Health Hazard 2 GHS P Statement
May intensify fire; oxidizer.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

Specifications

Chemical Name or Material Sodium hexanitrocobaltate(III)
CAS 13600-98-1
Molecular Formula CoN6Na3O12
Linear Formula Na3Co(NO2)6
Formula Weight 403.94
MDL Number MFCD00003512
Synonym Sodium cobaltinitrite
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Immerse in cool water/wrap in wet bandages. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
May intensify fire; oxidizer.
Suspected of causing cancer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause an allergic skin reaction.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.

SDS & Safety Documents