Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexanitritoiridate(III), 99%

$221.66 - $852.76

Chemical Identifiers

Chemical Name or Material Sodium hexanitritoiridate(III)
CAS 94022-51-2
Molecular Formula IrN6Na3O12
Linear Formula Na3Ir(NO2)6
Formula Weight 537.22
MDL Number MFCD00064621
Synonym Sodium hexakis(nitrito)iridate(III)
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2300202

 
alfa aesar™
B2300202
0.5g Each for $221.66

AAB2300288

 
alfa aesar™
B2300288
2.5g Each for $852.76
Description & Specifications

Specifications

Formula Weight 537.22
Chemical Name or Material Sodium hexanitritoiridate(III)
Assay 99%
Molecular Formula IrN6Na3O12
Linear Formula Na3Ir(NO2)6
MDL Number MFCD00064621
CAS 94022-51-2
Synonym Sodium hexakis(nitrito)iridate(III)
Quantity 0.5g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.