missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hexanitritocobaltate(III), ACS, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 01166736

 View more versions of this product

Catalog No. AA1166736



Description

Description

Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Sodium hexanitritocobaltate(III)
14649-73-1
Powder
CoN6Na3O12
UN1479
cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11
ZEBNDJQGEZXBCR-UHFFFAOYSA-H
trisodium;cobalt(3+);hexanitrite
159748
ACS
220°C (decomposition)
Yellow to Orange
500 g
MFCD00003512
14,8603
Very soluble in water
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3]
403.933
403.94
Odorless

Chemical Identifiers

14649-73-1
403.933
ZEBNDJQGEZXBCR-UHFFFAOYSA-H
159748
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3]
CoN6Na3O12
MFCD00003512
cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11
trisodium;cobalt(3+);hexanitrite
Safety and Handling

Safety and Handling

GHS H Statement
H272-H334-H351-H335-H315-H319-H317
May intensify fire; oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

P201-P202-P210-P220-P221-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P370+P378q-P501c

H272-H315-H317-H319-H334-H335-H350

DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.

EINECSNumber : 237-077-7

RTECSNumber : GF9480000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only