Learn More
Sodium hexanitritocobaltate(III), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1121630
Description
Sodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium hexanitritocobaltate(III) is used as primary and secondary intermediate in chemical research.
Solubility
Soluble in water.
Notes
Store at room temperature. Incompatible with acids, reducing agents and amines.
Specifications
Sodium hexanitritocobaltate (III) | |
14649-73-1 | |
CoN6Na3O12 | |
MFCD00003512 | |
14,8603 | |
Soluble in water. | |
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3] | |
403.933 | |
403.94 |
220°C (decomposition) | |
250 g | |
Na3Co(NO2)6 | |
UN1479 | |
cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11 | |
ZEBNDJQGEZXBCR-UHFFFAOYSA-H | |
trisodium;cobalt(3+);hexanitrite | |
159748 | |
Odorless |
Chemical Identifiers
14649-73-1 | |
403.933 | |
ZEBNDJQGEZXBCR-UHFFFAOYSA-H | |
159748 | |
N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].N(=O)[O-].[Na+].[Na+].[Na+].[Co+3] |
CoN6Na3O12 | |
MFCD00003512 | |
cobaltic trisodium hexanitrite, trisodium cobalt 3+ hexanitrite, sodium hexanitritocobaltate 3-, cobalt 3+ sodium nitrite 1:3:6, cobaltate 3-, hexanitrito-, trisodium, cobaltate 3-, hexakis nitrito-o-, trisodium, oc-6-11, na3 co no2 6, cobaltate 3-, hexakis nitrito-kappao-, sodium 1:3 , oc-6-11 | |
trisodium;cobalt(3+);hexanitrite |
Safety and Handling
GHS H Statement
H272-H334-H351-H335-H315-H319-H317
May intensify fire
oxidizer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Suspected of causing cancer.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
P201-P202-P210-P220-P221-P261-P264b-P271-P272-P280-P281-P285-P302+P352-P304+P340-P305+P351+P338-P308+P313-P333+P313-P342+P311-P362-P370+P378q-P501c
H272-H315-H317-H319-H334-H335-H350
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: OXIDIZING SOLID, N.O.S.
EINECSNumber : 237-077-7
RTECSNumber : GF9480000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only