missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexahydroxyplatinate(IV), 99.9% (metals basis)

Manufacturer:  Alfa Aesar™ 1262003

 View more versions of this product

Catalog No. AA1262003



Specifications

Specifications

Sodium hexahydroxyplatinate(IV)
150°C (decomposition)
12325-31-4
Na2Pt(OH)6
343.1
99.9%
Yellow to Brown
1g
(metals basis)
MFCD00014236
Crystalline

Chemical Identifiers

12325-31-4
MFCD00014236
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

RTECSNumber : TP1756000

TSCA : TSCA

Documents