Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexahydroxyplatinate(IV), 99.9% (metals basis)

Chemical Identifiers

Chemical Name or Material Sodium hexahydroxyplatinate(IV)
CAS 12325-31-4
Linear Formula Na2Pt(OH)6
Formula Weight 343.1
MDL Number MFCD00014236
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1262003

 
alfa aesar™
1262003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1262006

 
alfa aesar™
1262006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 343.1
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Chemical Name or Material Sodium hexahydroxyplatinate(IV)
TSCA TSCA
Physical Form Crystalline
Assay 99.9%
Assay Percent Notes (metals basis)
Linear Formula Na2Pt(OH)6
MDL Number MFCD00014236
CAS 12325-31-4
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Color Yellow to brown
Melting Point 150°C (dec.)
Quantity 1g
RTECS Number TP1756000
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see ... on this label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.