missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hexafluorosilicate, >99%, Alfa Aesar™

Manufacturer:  Alfa Aesar 69106A3

 View more versions of this product

Catalog No. AA69106A3



Specifications

Specifications

Sodium hexafluorosilicate
>99%
MFCD00003491
TWGUZEUZLCYTCG-UHFFFAOYSA-N
disodium;hexafluorosilicon(2-)
28127
Crystals
2kg
16893-85-9
F6Na2Si
sodium hexafluorosilicate, sodium fluosilicate, salufer, sodium silicofluoride, safsan, super prodan, destruxol applex, ortho earwig bait, ortho weevil bait, ens-zem weevil bait
F[Si-2](F)(F)(F)(F)F.[Na+].[Na+]
188.055
188.06
White

Chemical Identifiers

16893-85-9
188.055
TWGUZEUZLCYTCG-UHFFFAOYSA-N
28127
F[Si-2](F)(F)(F)(F)F.[Na+].[Na+]
F6Na2Si
MFCD00003491
sodium hexafluorosilicate, sodium fluosilicate, salufer, sodium silicofluoride, safsan, super prodan, destruxol applex, ortho earwig bait, ortho weevil bait, ens-zem weevil bait
disodium;hexafluorosilicon(2-)
Safety and Handling

Safety and Handling

H301-H311-H331: Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 240-934-8

SDS Documents