missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hexafluoroantimonate(V), 99.5%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1306622

 View more versions of this product

Catalog No. AA1306622



Specifications

Specifications

Sodium hexafluoroantimonate(V)
99.5%
MFCD00003482
sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium salt
F[Sb-](F)(F)(F)(F)F.[Na+]
258.74
258.73
3.38g/cm3
White
16925-25-0
F6NaSb
White powder
HKLMYZVMEYYVBS-UHFFFAOYSA-H
sodium;hexafluoroantimony(1-)
16689647
Powder
Odorless
100g

Chemical Identifiers

16925-25-0
258.74
HKLMYZVMEYYVBS-UHFFFAOYSA-H
16689647
F[Sb-](F)(F)(F)(F)F.[Na+]
F6NaSb
MFCD00003482
sodium hexafluoroantimonate, sodium hexafluoroantimonate v, sodium hexafluorostibanuide, sodium hexafluoro antimonate, antimony sodium fluoride, f6sb.na, nasbf6, acmc-20ajo7, sodium hexafluorostiboranuide, hexafluorostibine, sodium salt
sodium;hexafluoroantimony(1-)
Safety and Handling

Safety and Handling

H302-H332-H411: Harmful if swallowed. Harmful if inhaled. Toxic to aquatic life with long lasting effects.

DOTInformation : Hazard Class: 6.1, Packing Group: III

EINECSNumber : 240-989-8

Documents