missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium hexafluoroaluminate, 99.9% (metals basis), Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific 4447836

 View more versions of this product

Catalog No. AA4447836



Specifications

Specifications

Sodium hexafluoroaluminate
99.9% (metals basis)
MFCD00003507
209.95
2.9
White
500g
13775-53-6
Na3AlF6
Soluble in water(0.4g/L).
Powder
Odorless
1000°C

Chemical Identifiers

13775-53-6
Na3AlF6
MFCD00003507
Safety and Handling

Safety and Handling

Do not breathe dust. Wear suitable gloves. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). Avoid release to the environment. Refer to special instructions/Safety data sheets.

P260-P264b-P270-P271-P304+P340-P312-P314-P501c

H332-H372

DOTInformation : DOT Class: 9; Packaging Group: III

EINECSNumber : 237-410-6

RTECSNumber : BD0075000

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents