Learn More
Sodium hexafluoroaluminate, 99.4% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 03634530
Description
Mainly used as fluxing agent in the production of aluminum electrolysis, also used as additive to the wear-resisting of abrasive products, iron alloy and the flux of rimming steel, non-ferrous metal flux, casting deoxidizer, enamel of emulsifier, glass opal agent, welding flux and ceramics filler. Aluminum- metallurgy. This chemical is used as adhesives, cosolvent in enamel industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hexafluoroaluminate | |
13775-53-6 | |
250 g | |
MFCD00003507 | |
Aluminum sodium fluoride; Cryolite | |
209.95 | |
Odorless |
1,000°C | |
Lump | |
(metals basis) | |
UN3260 | |
Sparingly soluble in water 0.4g/L @ 20°C. | |
99.5% | |
2.90 g/mL |
Chemical Identifiers
13775-53-6 | |
MFCD00003507 | |
Aluminum sodium fluoride; Cryolite |
Safety and Handling
GHS H Statement
H372-H302-H332
Causes damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
P260-P264b-P270-P271-P304+P340-P312-P314-P501c
H332-H372
DOTInformation : Hazard Class: 9; Packaging Group: III
EINECSNumber : 237-410-6
RTECSNumber : BD0075000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only