missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexafluoroaluminate, 97.8+%

Manufacturer:  Alfa Aesar™ 8827536

 View more versions of this product

Catalog No. AA8827536



Specifications

Specifications

Sodium hexafluoroaluminate
Na3AlF6
UN3260
209.95
≥97.8%
Odorless
500g
13775-53-6
MFCD00003507
Cryolite; Aluminum sodium fluoride
Powder
2.90g/mL
1,000°C

Chemical Identifiers

13775-53-6
Na3AlF6
MFCD00003507
Cryolite; Aluminum sodium fluoride
Safety and Handling

Safety and Handling

GHS H Statement
H372-H302-H332
Causes damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
P260-P261-P304+P340-P301+P312-P312-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

RTECSNumber : BD0075000

TSCA : TSCA

SDS Documents