Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexachlororhodate(III) dodecahydrate, Rh 17.1%

Chemical Identifiers

Chemical Name or Material Sodium hexachlororhodate(III) dodecahydrate
CAS 14972-70-4
Formula Weight 600.77 (384.59 Anhydrous)
MDL Number MFCD00003509
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1180903

  SDS
alfa aesar™
1180903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1180906

  SDS
alfa aesar™
1180906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 600.77 (384.59 Anhydrous)
Chemical Name or Material Sodium hexachlororhodate(III) dodecahydrate
TSCA TSCA
Physical Form Crystalline
MDL Number MFCD00003509
CAS 14972-70-4
Melting Point 900°C (dec.)
Concentration Rh 17.1%
Quantity 1g