Learn More
Sodium hexachloropalladate(IV) hydrate, 99.9% (metals basis), Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01103906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hexachloropalladate(IV) hydrate | |
Brown to Red | |
Cl6Na2Pd | |
MFCD00014235 | |
Hygroscopic | |
[Na+].[Na+].Cl[Pd++](Cl)(Cl)(Cl)(Cl)Cl | |
365.10 | |
99.9% |
53823-60-2 | |
(metals basis) | |
Na2PdCl6·xH2O | |
5 g | |
OWJQEDIJRBKDJO-UHFFFAOYSA-H | |
disodium hexachloropalladiumbis(ylium) | |
365.10 (Anhydrous) | |
Crystalline |
Chemical Identifiers
53823-60-2 | |
365.10 | |
OWJQEDIJRBKDJO-UHFFFAOYSA-H | |
[Na+].[Na+].Cl[Pd++](Cl)(Cl)(Cl)(Cl)Cl |
Cl6Na2Pd | |
MFCD00014235 | |
disodium hexachloropalladiumbis(ylium) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 258-809-1
TSCA : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only