missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min

Manufacturer:  Alfa Aesar™ 1217601

 View more versions of this product

Catalog No. AA1217601



Specifications

Specifications

Sodium hexachloroosmate(IV) dihydrate
0.25g
Cl6Na2Os
MFCD00014234
PVTVRPSPEDISMV-UHFFFAOYSA-H
disodium;hexachloroosmium(2-)
44151190
Crystalline
Odorless
Brown to Red
1307-81-9
Na2OsCl6·2H2O
Os 38.7% min.
[Na+].[Na+].Cl[Os-2](Cl)(Cl)(Cl)(Cl)Cl
448.91
484.93 (448.90 Anhydrous)
Os 38.7% min.
Air & Moisture Sensitive

Chemical Identifiers

1307-81-9
448.91
PVTVRPSPEDISMV-UHFFFAOYSA-H
disodium;hexachloroosmium(2-)
Cl6Na2Os
MFCD00014234
44151190
[Na+].[Na+].Cl[Os-2](Cl)(Cl)(Cl)(Cl)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents