Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min

Chemical Identifiers

Chemical Name or Material Sodium hexachloroosmate(IV) dihydrate
CAS 1307-81-9
Linear Formula Na2OsCl6·2H2O
Formula Weight 484.93 (448.90 Anhydrous)
MDL Number MFCD00014234
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1217601

  SDS
alfa aesar™
1217601
0.25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1217603

  SDS
alfa aesar™
1217603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1217606

  SDS
alfa aesar™
1217606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 484.93 (448.90 Anhydrous)
Chemical Name or Material Sodium hexachloroosmate(IV) dihydrate
Physical Form Crystalline
Linear Formula Na2OsCl6·2H2O
MDL Number MFCD00014234
CAS 1307-81-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Red to brown
Concentration Os 38.7% min.
Quantity 0.25g
Odor Odorless
Sensitivity Air & Moisture Sensitive
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.