Learn More
Sodium hexachloroosmate(IV) dihydrate, Os 38.7% min, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01217603
Description
Sodium hexachloroosmate(IV) dihydrate is used as an intermediate and catalyst in organic synthesis. It is also used to prepare dihydrogen complexes of osmium(II) containing meso-tetraphos-1 ligand.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium hexachloroosmate(IV) dihydrate | |
1307-81-9 | |
1 g | |
Na2OsCl6·2H2O | |
Insoluble in water. | |
[Na+].[Na+].Cl[Os--](Cl)(Cl)(Cl)(Cl)Cl | |
448.91 | |
484.93 (448.90 Anhydrous) | |
Odorless |
Brown to Red | |
Crystalline | |
Cl6Na2Os | |
MFCD00014234 | |
PVTVRPSPEDISMV-UHFFFAOYSA-H | |
disodium;hexachloroosmium(2-) | |
44151190 | |
Os 38.7% min. | |
Air & Moisture Sensitive |
Chemical Identifiers
1307-81-9 | |
448.91 | |
PVTVRPSPEDISMV-UHFFFAOYSA-H | |
[Na+].[Na+].Cl[Os--](Cl)(Cl)(Cl)(Cl)Cl |
Cl6Na2Os | |
MFCD00014234 | |
44151190 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
EINECSNumber : 215-152-5
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only