Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexachloroiridate(IV) hexahydrate, Ir 33.9% min

Chemical Identifiers

Chemical Name or Material Sodium hexachloroiridate(IV) hexahydrate
CAS 19567-78-3
Formula Weight 558.99 (450.92 Anhydrous)
MDL Number MFCD00149169
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1189003

 
alfa aesar™
1189003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1189006

 
alfa aesar™
1189006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 558.99 (450.92 Anhydrous)
Chemical Name or Material Sodium hexachloroiridate(IV) hexahydrate
TSCA TSCA
Physical Form Powder
MDL Number MFCD00149169
CAS 19567-78-3
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H290-H302-H319
May be corrosive to metals.
Harmful if swallowed.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P301+P312-P337+P313-P406-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Color Red-black
Melting Point 600°C (dec.)
Concentration Ir 33.9% min.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P301+P312-P337+P313-P406-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
If eye irritation persists: Get medical advice/attention.
Store in corrosive resistant container with a resistant inner liner.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H290-H302-H319
May be corrosive to metals.
Harmful if swallowed.
Causes serious eye irritation.