missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexachloroiridate(III) hydrate, Ir 31.5% min

Manufacturer:  Alfa Aesar™ 1188903

 View more versions of this product

Catalog No. AA1188903



Specifications

Specifications

Sodium hexachloroiridate(III) hydrate
1g
Cl6H2IrNa3O
sodium hexachloroiridate iii hydrate, sodium hexachloroiridate 3--water 3/1/1, cl6ir.3na.h2o, sodium hexachloroiridate iii hydrate, ir, iridate 3-,hexachloro-, sodium, hydrate 1:3:? , oc-6-11
DXIIVDVTQZORNM-UHFFFAOYSA-H
trisodium;hexachloroiridium(3-);hydrate
56845692
Crystalline
Moisture Sensitive
123334-23-6
MFCD00003508
Ir 31.5% min.
O.[Na+].[Na+].[Na+].Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl
491.901
473.89 (Anhydrous)
Ir 31.5% min.

Chemical Identifiers

123334-23-6
491.901
DXIIVDVTQZORNM-UHFFFAOYSA-H
56845692
O.[Na+].[Na+].[Na+].Cl[Ir-3](Cl)(Cl)(Cl)(Cl)Cl
Cl6H2IrNa3O
MFCD00003508
sodium hexachloroiridate iii hydrate, sodium hexachloroiridate 3--water 3/1/1, cl6ir.3na.h2o, sodium hexachloroiridate iii hydrate, ir, iridate 3-,hexachloro-, sodium, hydrate 1:3:? , oc-6-11
trisodium;hexachloroiridium(3-);hydrate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents