Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexachloroiridate(III) hydrate, Ir 31.5% min

Chemical Identifiers

Chemical Name or Material Sodium hexachloroiridate(III) hydrate
Formula Weight 473.89 (Anhydrous)
MDL Number MFCD00003508
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1188903

 
alfa aesar™
1188903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA1188906

 
alfa aesar™
1188906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 473.89 (Anhydrous)
Chemical Name or Material Sodium hexachloroiridate(III) hydrate
TSCA TSCA
Physical Form Crystalline
MDL Number MFCD00003508
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration Ir 31.5% min.
Quantity 1g
Sensitivity Moisture Sensitive
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.