Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexabromorhodate(III) hydrate, Premion™, 99.99% (metals basis), Rh 11.4% min

Chemical Identifiers

Chemical Name or Material Sodium hexabromorhodate(III) hydrate
Purity Grade Premion™
Formula Weight 384.59 (Anhydrous)
MDL Number MFCD00798531
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4040803

  SDS
alfa aesar™
4040803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4040806

  SDS
alfa aesar™
4040806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 384.59 (Anhydrous)
Physical Form Crystalline Powder
Purity Grade Premion™
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
MDL Number MFCD00798531
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration Rh 11.4% min.
Assay Percent Notes (metals basis)
Assay 99.99%
Chemical Name or Material Sodium hexabromorhodate(III) hydrate
Health Hazard 1 Warning
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.