Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium hexabromoosmate(IV), Premion™, 99.95% (metals basis), Os 26.1% min

Chemical Identifiers

Chemical Name or Material Sodium hexabromoosmate(IV)
Purity Grade Premion™
Linear Formula Na2OsBr6
Formula Weight 715.64
MDL Number MFCD00798532
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4041203

  SDS
alfa aesar™
4041203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4041206

  SDS
alfa aesar™
4041206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Formula Weight 715.64
Chemical Name or Material Sodium hexabromoosmate(IV)
Purity Grade Premion™
Assay 99.95%
Assay Percent Notes (metals basis)
Linear Formula Na2OsBr6
MDL Number MFCD00798532
Color Red to black
Quantity 1g
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Concentration Os 26.1% min.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.