missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium glycolate, 97%

Manufacturer:  Alfa Aesar A1234122

 View more versions of this product

Catalog No. AAA1234122


 

Description

Description

Applications
Sodium glycolate is used as a disintegrant, suspending agent, gelling agent, buffering agent and in pharmaceutical grade for tablets and capsules. Further, it is used in cosmetics and personal care products primarily as an exfoliant, as pH adjusters, skin-conditioning agents and as a flavoring agent.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Specifications

Sodium Glycolate
97%
C2H3NaO3
MFCD00065358
C(C(=O)[O-])O.[Na+]
98.033
98.03
100g
2836-32-0
97%
HOCH2CO2Na
VILMUCRZVVVJCA-UHFFFAOYSA-M
sodium;2-hydroxyacetate
517347
Lump/Powder

Chemical Identifiers

2836-32-0
98.033
VILMUCRZVVVJCA-UHFFFAOYSA-M
C2H3NaO3
MFCD00065358
517347
Safety and Handling

Safety and Handling

RTECSNumber : MC5525000

TSCA : TSCA

SDS Documents