Please login to your online account to display your discounted pricing

Sodium glycolate, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  351570250

 View more versions of this product

Catalog No. AC351570250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Powder
Formula Weight 98.03
Quantity 25g
Color White to off-white
TSCA TSCA
RTECS Number MC5525000
EINECS Number 220-624-9
Synonym Sodium hydroxyacetate
Packaging Glass bottle
Infrared Spectrum Authentic
Assay Percent Range 97%
Linear Formula NaOOCCHOH
MDL Number MFCD00065358
Name Note 97%
Molecular Formula C2H3NaO3
CAS 2836-32-0
Chemical Name or Material Sodium glycolate

Specifications

Chemical Name or Material Sodium glycolate
Name Note 97%
CAS 2836-32-0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C2H3NaO3
Linear Formula NaOOCCHOH
Formula Weight 98.03
MDL Number MFCD00065358
Synonym Sodium hydroxyacetate