missing translation for 'onlineSavingsMsg'
Learn More

Sodium fumarate, 98%

CAS: 17013-01-3 | C4H4Na2O4 | 162.052 g/mol

Supplier:  Thermo Scientific Chemicals A112760E

 View more versions of this product

Catalog No. AAA112760E


Only null left
Explore available promotions

Description

Description

Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Sodium fumarate is used as an acidity regulator in food processing and as terminal electron acceptors in the cultivation of certain anaerobic microorganisms. It is also used as a buffering agent and pH adjuster in cosmetic and personal care products. Further, it is used as a food additive.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong oxidizing agents and strong acids.
Specifications

Specifications

Sodium fumarate
98%
MFCD00064567
Hygroscopic
Soluble in water.
C(=CC(=O)O)C(=O)O.[Na].[Na]
162.052
160.04
17013-01-3
C4H4Na2O4
2500 g
disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
VXXVUHAXJHEYFH-SEPHDYHBSA-N
(E)-but-2-enedioic acid;sodium
87110070
98%

Chemical Identifiers

17013-01-3
162.052
VXXVUHAXJHEYFH-SEPHDYHBSA-N
87110070
C(=CC(=O)O)C(=O)O.[Na].[Na]
C4H4Na2O4
MFCD00064567
disodium fumarate, sodium fumarate, sodium fumarate dibasic, fumaric acid, disodium salt, fumaric acid disodium salt, fumaran sodny czech, fumaran sodny, disodium butenedioate, di-sodium fumarate, sodium monofumarate
(E)-but-2-enedioic acid;sodium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 241-087-7

RTECSNumber : LT1830000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only