Please login to your online account to display your discounted pricing

Sodium fumarate, 98%, ACROS Organics™


Quantity 2.5kg
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC215530025

 View more versions of this product

Catalog No. AC215530025

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Powder
Assay ≥97.5%
Formula Weight 160.04
Quantity 2.5kg
Color White
Chemical Name or Material Sodium fumarate
Assay Percent Range 98%
Infrared Spectrum Authentic
Molecular Formula C4H2Na2O4
Linear Formula NaO2CCH=CHCO2Na
MDL Number MFCD00064567
CAS 17013-01-3
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Sodium fumarate
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C4H2Na2O4
Linear Formula NaO2CCH=CHCO2Na
Formula Weight 160.04
MDL Number MFCD00064567
Accessories

GHS P Statement:
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.