Please login to your online account to display your discounted pricing

Sodium Formate, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$810.98 - $810.98

Chemical Identifiers

Chemical Name or Material Sodium Formate
CAS 141-53-7
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula HCOONa
Formula Weight 68.01
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004093

 
avantor performance
370005
Poly Bottle 2.5kg Each for $810.98
Description & Specifications

Specifications

Physical Form Crystals
Formula Weight 68.01
Purity Grade Baker Analyzed™ ACS Reagent
Chemical Name or Material Sodium Formate
CAS 141-53-7
Molecular Formula HCOONa
Quantity 2.5kg
Specific Gravity 1L = 1.92kg
Packaging Poly Bottle