Learn More
Sodium formaldehydesulfoxylate dihydrate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 149190050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Reduction reagentSpecifications
Sodium formaldehydesulfoxylate dihydrate | |
97.5% min. (Iodometry) | |
CH3NaO3S·2H2O | |
MFCD00150598 | |
15,8755 | |
Solubility in water: 1088g/L. Other solubilities: practically insoluble in abs.alcohol, ether and, benzene | |
C(O)S(=O)[O-].O.O.[Na+] | |
154.11 | |
154.11 |
6035-47-8 | |
Plastic bottle | |
HOCH2SO2Na·2H2O | |
5kg | |
sodium hydroxymethanesulfinate dihydrate, sodium formaldehydesulfoxylate dihydrate, methanesulfinic acid, hydroxy-, monosodium salt, dihydrate, sodium formaldehyde sulfoxylate, sodium dihydrate hydroxymethanesulfinate, sodium formaldehyde sulfoxylate nf, formaldehyde sodium sulfoxylate dihydrate, monosodium hydroxymethanesulfinate dihydrate, 79-25-4 parent, hydroxymethanesulfinic acid monosodium salt dihydrate | |
UCWBKJOCRGQBNW-UHFFFAOYSA-M | |
sodium;hydroxymethanesulfinate;dihydrate | |
23666330 | |
98% |
Safety and Handling
GHS H Statement
Suspected of causing genetic defects.
Suspected of damaging the unborn child.
Contact with acids liberates very toxic gas.
GHS P Statement
Obtain special instructions before use.
Wear protective gloves/protective clothing/eye protection/face protection.
IF exposed or concerned: Get medical advice/attention.
GHS Signal Word: Warning
RUO â Research Use Only