Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium fluorophosphate

$61.42 - $61.42

Chemical Identifiers

Chemical Name or Material Sodium fluorophosphate
CAS 10163-15-2
Formula Weight 143.95
MDL Number MFCD00014248
UN Number UN3288
Synonym Disodium fluorophosphate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA1412936

 
alfa aesar™
1412936
500g Each for $61.42
Description & Specifications

Specifications

Formula Weight 143.95
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Sodium fluorophosphate
Odor Odorless
Physical Form Powder
MDL Number MFCD00014248
CAS 10163-15-2
UN Number UN3288
Synonym Disodium fluorophosphate
Quantity 500g
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.