Please login to your online account to display your discounted pricing

Sodium Fluoride, Powder, U.S.P., J.T.Baker™

$514.00 - $2,081.26

Chemical Identifiers

Chemical Name or Material Sodium Fluoride
CAS 7681-49-4
Purity Grade USP
Molecular Formula NaF
Formula Weight 41.99
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02004089

 
avantor performance
368905
Poly Bottle 2kg Each for $2,081.26

02004091

 
avantor performance
368901
Poly Bottle 500g Each for $514.00
Description & Specifications

Specifications

Formula Weight 41.99
CAS 7681-49-4
Chemical Name or Material Sodium Fluoride
Molecular Formula NaF
Physical Form Powder
Purity Grade USP
Specific Gravity 1L = 2.78kg
Packaging Poly Bottle
Quantity 2kg