missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium fluoride, ACS, 99% min. , Alfa Aesar™

Steel Degreasing Agent, in Tin Electroplating, Flux

Manufacturer:  Alfa Aesar 1156130

 View more versions of this product

Catalog No. AA1156130



Specifications

Specifications

Sodium fluoride
7681-49-4
FNa
White powder
PUZPDOWCWNUUKD-UHFFFAOYSA-M
sodium;fluoride
5235
41.99
ACS
Odorless
White
For ACS analysis
99% min.
MFCD00003524
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium flouride, antibulit, floridine, fluonatril
[F-].[Na+]
41.988
CHEBI:28741
Powder
2.78g/cm3 at 20°C
1.3 hPa (1mm Hg)
250g

Chemical Identifiers

7681-49-4
41.988
PUZPDOWCWNUUKD-UHFFFAOYSA-M
5235
sodium;fluoride
FNa
MFCD00003524
sodium fluoride, florocid, fluoride, sodium, zymafluor, ossin, sodium fluoride naf, sodium flouride, antibulit, floridine, fluonatril
CHEBI:28741
[F-].[Na+]
Safety and Handling

Safety and Handling

H301-H315-H319: Toxic if swallowed. Causes skin irritation. Causes serious eye irritation.

DOTInformation : DOT Class: 6.1, Packaging Group: III

EINECSNumber : 231-667-8

Documents